Learn More
Fisherbrand™ Traceable™ Data Acquisition Software
Product Code. 17156324 Shop All Cole-Parmer Products
Change view
Click to view available options
Includes:
Cailbration, Software CD and 6 ft. Cables (USB, Serial, and Instrument Connections)
Software CD and 6 ft. Cables (USB, Serial, and Instrument Connections)
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
17156324 Cailbration, Software CD and 6 ft. Cables (USB, Serial, and Instrument Connections) Each
17110089 Software CD and 6 ft. Cables (USB, Serial, and Instrument Connections) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17156324 Supplier Fisherbrand™ Supplier No. 9008021
  • ONLINE EXCLUSIVE PRICE
    532.00€
    465.49€ / paquete de 1
    Ahorro 66.51€
    12% Descuento
Fecha estimada de envoi: 25-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Complete Data Acquisition Software enables measurement readings to be displayed and stored on a PC from up to 8 devices at a time.
  • Display and store measurement readings on a PC from up to 8 devices at a time
  • Data capture runs in the background, enabling users to work on other programs simultaneously
  • Set audible and visual notification alarms, or immediately send an email if an over/under condition occurs
  • Software can be used to connect a meter directly to PC via cable
  • Check software specification tables for meter compatibility
Specifications
Type Data Acquisition Software
Display Type No Display
Description Data Acquisition Software with USB Cable and Calibration
Includes Cailbration, Software CD and 6 ft. Cables (USB, Serial, and Instrument Connections)
For Use With (Equipment) 90080-09, 03313-65, 03313-66, 37804-06, 37804-07 (Traceable™ 4000, 4080, 4085, 6412, 6413)