Learn More
Novus Biologicals™ DARPP-32 Overexpression Lysate
Marca:  Novus Biologicals™ NBL1-14679
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18216265
  • 685.73€ / 0.10 miligramo
Fecha estimada de envío: 21-08-2024
para ver el stock
Descripción
Descripción
The DARPP-32 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Especificaciones
Especificaciones
Western Blot
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
The lysate was created in HEK293T cells, using plasmid ID RC212689 and based on accession number NM_32192. The protein contains a C-terminal DDK tag.
RIPA buffer
Homo sapiens protein phosphatase 1, regulatory (inhibitor) subunit 1B (PPP1R1B), transcript variant 1, mRNA.
Native