missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAPK1 Recombinant Human Protein
DAPK1 Recombinant Human Protein
Marca: Invitrogen PV3969
Detalles adicionales : Peso : 0.00900kg
Descripción
DAPK1 is a large, calmodulin-dependent kinase involved in gamma-interferon induced programmed cell death and is a potential tumour suppressor.Especificación
NP_004929 | |
Cell viability, proliferation and function, pharma and biopharma, protein assays and analysis, target and lead identification and validation | |
71.6kDa | |
85% by SDS-PAGE | |
–80°C | |
1 Each | |
DAPK1 | |
DAPK1 was pre-diluted in enzyme dilution buffer and assayed in 12.5mM Tris (pH 7.5), 10mM MgCl2, 1mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2mM DTT, 0.01% Triton™ X-100, 100μM ATP, 200μM DAPKtide peptide substrate (KKRPQRRYSNVF) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
Kinase and inhibitors | |
DAPK | |
DAPK1 Recombinant Human Protein | |
Human |
0.31mg/mL | |
1612, 7124 | |
DAPK1 Recombinant Human Protein | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
Serine/threonine kinase | |
DAPK1 Recombinant Human Protein | |
Kinase assay | |
Baculovirus | |
Amino acids 1-363 | |
60nmoles of phosphate transferred to DAPKtide peptide substrate (KKRPQRRYSNVF) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 3.33μg/mL | |
GST Tag |
For Research Use Only. Not for use in diagnostic procedures.