missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAPK1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25136-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Death-associated protein kinase 1 is a positive mediator of gamma-interferon induced programmed cell death. DAPK1 encodes a structurally unique 160-kD calmodulin dependent serine-threonine kinase that carries 8 ankyrin repeats and 2 putative P-loop consensus sites. It is a tumor suppressor candidate.Especificaciones
DAPK1 | |
Polyclonal | |
Unconjugated | |
DAPK1 | |
DAP kinase 1, DAPK, DAPK1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1612 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1.01 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P53355 | |
DAPK1 | |
DAPK1 Fusion Protein Ag18387 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |