missing translation for 'onlineSavingsMsg'
Learn More

DAP5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 17728-1-AP-20UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16817684

  • 121.38€ / 20 microlitros
Envío estimado: 13-12-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

CDK5RAP2 is a regulator of CDK5 (cyclin-dependent kinase 5) activity. The protein is localized to the centrosome and Golgi complex and interacts with CDK5R1 and pericentrin (PCNT). CDK5RAP2 plays a role in centriole engagement and microtubule nucleation, and has been linked to primary microcephaly and Alzheimer′s disease. Alternative splicing results in multiple transcript variants.
Especificaciones

Especificaciones

DAP5
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P78344, Q62448
AAG1, DAP 5, DAP5, Death associated protein 5, eIF 4 gamma 2, eIF 4G 2, eIF4G 2, EIF4G2, NAT1, p97
Rabbit
IgG
20 μL
-20°C
Primary
13690, 1982, 361628
Western Blot, Immunoprecipitation
Unconjugated
EIF4G2
Liquid
Eif4g2
DAP5 Fusion Protein Ag12125
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Mouse, Human, Rat
Videos
SDS
Documentos
Certificados
Ofertas especiales

Ofertas especiales