missing translation for 'onlineSavingsMsg'
Learn More

DAP3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10276-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16806563

  • 399.00€ / 150 microlitros
Fecha estimada de envío: 17-04-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that also participates in apoptotic pathways which are initiated by tumor necrosis factor-alpha, Fas ligand, and gamma interferon. This protein potentially binds ATP/GTP and might be a functional partner of the mitoribosomal protein S27. Splice variants that differ in the 5′ UTR have been found for this gene; both variants encode the same protein. Pseudogenes corresponding to this gene are found on chromosomes 1q and 2q.
Especificaciones

Especificaciones

DAP3
Polyclonal
Unconjugated
DAP3
bMRP 10, DAP 3, DAP3, death associated protein 3, MRP S29, MRPS29, S29mt
Rabbit
Antigen Affinity Chromatography
RUO
65111, 7818
-20°C
Liquid
Western Blot
0.26 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P51398, Q9ER88
Dap3
DAP3 Fusion Protein Ag0274
150 μL
Primary
Human, Mouse
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales