missing translation for 'onlineSavingsMsg'
Learn More

Sal sódica de D-mio-inositol-1,2,6-trifosfato, 98 %, Thermo Scientific Chemicals

Catalog Number 15450947
Click to view available options
Cantidad:
0.1 mg
0.5 mg
Este artículo no se puede devolver. Vea la política de devoluciones
MFCD12912249
1,2,6-IP3; Ins(1,2,6)-P3 sodium salt
Este artículo no se puede devolver. Vea la política de devoluciones
MFCD12912249
1,2,6-IP3; Ins(1,2,6)-P3 sodium salt

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material D-myo-Inositol-1,2,6-trisphosphate sodium salt
Color Blanco
Cantidad 0.1 mg
Formula Weight (peso de la fórmula) 486.04
Porcentaje de pureza 98%
Forma física Polvo liofilizado

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'