Learn More
D-Mannosamine Hydrochloride, TRC
Product Code. 30010652
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010652 100 mg 100mg
30012432 1 g 1g
30004998 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010652 Supplier Toronto Research Chemicals Supplier No. TRCM167000100MG
 Restricted Item
Fecha estimada de envoi: 17-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5505-63-5
Molecular Formula C6 H13 N O5 . Cl H
Molecular Weight (g/mol) 215.63
IUPAC Name (2S,3R,4S,5R)-2-amino-3,4,5,6-tetrahydroxyhexanal;hydrochloride
SMILES Cl.N[C@H](C=O)[C@@H](O)[C@H](O)[C@H](O)CO
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C6H13NO5.ClH/c7-3(1-8)5(11)6(12)4(10)2-9;/h1,3-6,9-12H,2,7H2;1H/t3-,4-,5-,6-;/m1./s1
Formula Weight 215.0561
Chemical Name or Material D-Mannosamine hydrochloride