missing translation for 'onlineSavingsMsg'
Learn More

D-Dimer Antibody (DD44), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NB100-73039B

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215275

  • 402.80€ / 0.20 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

D-Dimer Monoclonal specifically detects D-Dimer in Human samples. It is validated for Western Blot, ELISA.
Especificaciones

Especificaciones

D-Dimer
Monoclonal
Biotin
PBS with 0.05% Sodium Azide
Mouse
Protein A purified
RUO
D-dimer and high molecular weight fibrin degradation products, no cross-reactivity with fibrinogen
Store at 4C in the dark.
IgG2b
Western Blot, ELISA
DD44
Western Blot, ELISA
D Dimer
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with mixture of D-dimer and high molecular weight fibrin degradation products.
0.2 mL
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales