Learn More
Epredia™ Cytospin™ 4 Cytofunnel™ Starter Kit
Product Code. 12097139 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Clipped Funnel Starter Kit
EZ Cytofunnel Starter Kit
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
12097139 EZ Cytofunnel Starter Kit Each
12017159 Clipped Funnel Starter Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12097139 Supplier Epredia™ Supplier No. A78710013
  • ONLINE EXCLUSIVE PRICE
    792.00€
    664.03€ / paquete de 1
    Ahorro 127.97€
    16% Descuento
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Accessory Starter Kit for the Epredia™ Cytospin™ 4 Cytocentrifuge. (Centrifuge is sold separately, Cat. No. A78300003)
The Cytofunnel Starter Kit provides everything needed for new Cytospin 4 users.
Available in both standard clipped Cytofunnel and EZ Cytofunnel options:
  • Clipped Cytofunnel Starter Kit
    • Includes: 50 Single White Cytofunnels, 25 Double Cytofunnels, Cytoslides, 12 Cytoclips
  • EZ Cytofunnel Starter Kit
    • Includes: 25 EZ Megafunnels, 20 EZ Single White Cytofunnels, 20 EZ Double Cytofunnels and Cytoslides
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Cytospin™ 4 Cytocentrifuge
Product Line Cytospin
Frequency 50/60 Hz
Voltage 100/240 V
Product Type EZ Cytofunnel Starter Kit