missing translation for 'onlineSavingsMsg'
Learn More

Cytosolic beta-Glucosidase/GBA3 Antibody (008), PE, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Bio-Techne NBP2-89924PE

Additional Details : Peso : 0.00970kg

Product Code. 18215358

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Description

Description

Cytosolic beta-Glucosidase/GBA3 Monoclonal specifically detects Cytosolic beta-Glucosidase/GBA3 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Cytosolic beta-Glucosidase/GBA3
Monoclonal
PE
PBS
Rabbit
Protein A purified
RUO
Primary
Human
Purified
Flow Cytometry
8
Flow Cytometry
CBG, CBGL1GLUC, cytosolic beta-glucosidase, Cytosolic beta-glucosidase-like protein 1, EC 3.2.1.21, glucosidase, beta, acid 3 (cytosolic), klotho-related protein, KLrP, MGC104276, MGC126878
This antibody was obtained from a rabbit immunized with purified, recombinant Human Cytosolic beta-Glucosidase/GBA3 (NP_066024.1; Met1-Leu469).
0.1 mL
metabolism
57733
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers