missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 8/18 Mouse, Clone: KRT8.18/1346, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio MSM27-1346-P1
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Cytokeratin 8/18 | |
Cytoplasmic | |
KRT8.18/1346 | |
Unconjugated | |
Mouse | |
MCF-7 or A431 cells. Skin, Colon, lung or breast carcinoma | |
100 μg | |
Primary | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
Cytoskeleton preparation from HeLa cells | |
Protein A/G Purified | |
RUO | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |