missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 3 Antibody (KRT3/2130), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP2-79852JF549

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213328

  • 578.82€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Cytokeratin 3 Monoclonal specifically detects Cytokeratin 3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

Cytokeratin 3
Monoclonal
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
KRT3
Recombinant full-length human Cytokeratin 3 protein (Uniprot: P12035)
0.1 mL
Primary
Store at 4°C in the dark.
Immunohistochemistry, Immunohistochemistry (Paraffin)
KRT3/2130
Immunohistochemistry, Immunohistochemistry-Paraffin
CK3, CK-3, cytokeratin-3, FLJ95909, K3cytokeratin 3, keratin 3, keratin, type II cytoskeletal 3,65 kDa cytokeratin, Keratin-3, Type-II keratin Kb3
Mouse
Protein A or G purified
RUO
Human
IgG1 κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales