missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 19 Antibody (OTI3F8), Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP1-48277

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213442

  • 572.34€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Cytokeratin 19 Monoclonal specifically detects Cytokeratin 19 in Human, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

Cytokeratin 19
Monoclonal
0.15 mg/mL
Western Blot 1:1000, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50
CK19, CK-19, cytokeratin 19, Cytokeratin-19,40-kDa keratin intermediate filament, K19cytokeratin-19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, keratin-19, MGC15366
Mouse
43.9 kDa
0.1 mL
Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Stem Cell Markers
3880
Human, Rat, Canine, Primate, Monkey
IgG1
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
OTI3F8
Unconjugated
P08727
KRT19
Recombinant fragment expressed in E.coli corresponding to amino acids 240-390 of human CK19
Affinity Purified
RUO
Primary
This antibody is specific for Homo sapiens keratin 19 (KRT19).
Store at -20°C. Avoid freeze-thaw cycles.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only