missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 19 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NB100-687AF594

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213761

  • 561.54€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

Cytokeratin 19 Polyclonal specifically detects Cytokeratin 19 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

Cytokeratin 19
Polyclonal
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
CK19, CK-19, cytokeratin 19, Cytokeratin-19,40-kDa keratin intermediate filament, K19cytokeratin-19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, keratin-19, MGC15366
Rabbit
0.1 mL
3880
Store at 4°C in the dark.
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
KRT19
Synthetic peptide derived from the C-terminal region of human Cytokeratin 19 (between residues 350-400) [UniProt P08727]
Primary
Human, Mouse, Rat
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales