missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 15 Antibody (SPM190), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP2-54453DL594

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18216094

  • 569.10€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

Cytokeratin 15 Monoclonal specifically detects Cytokeratin 15 in Human, Rat, Bovine samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Especificaciones

Especificaciones

Cytokeratin 15
Monoclonal
DyLight 594
50mM Sodium Borate with 0.05% Sodium Azide
KRT15
A 17 amino acids synthetic peptide from the C- terminal of Human Cytokeratin 15 (Uniprot: P19012)
0.1 mL
Cell Biology, Cellular Markers
Human, Rat, Bovine
IgG2a κ
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
SPM190
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
CK15, CK-15, cytokeratin 15, cytokeratin-15, K15keratin-15, basic, K1CO, keratin 15, keratin, type I cytoskeletal 15, Keratin-15, keratin-15, beta, KRTB, type I cytoskeletal 15
Mouse
Protein A or G purified
RUO
Primary
Store at 4°C in the dark.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales