missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin F Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12073-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins and the kininogens. The type 2 cystatin proteins are a class of cysteine proteinase inhibitors found in a variety of human fluids and secretions. This gene encodes a glycosylated cysteine protease inhibitor with a putative role in immune regulation through inhibition of a unique target in the hematopoietic system. Expression of the protein has been observed in various human cancer cell lines established from maligt tumors.Especificaciones
Cystatin F | |
Polyclonal | |
Unconjugated | |
CST7 | |
CMAP, CST7, Cystatin 7, Cystatin F, cystatin F (leukocystatin), Leukocystatin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13011, 8530 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O76096, O89098 | |
CST7 | |
Cystatin F Fusion Protein Ag2697 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |