missing translation for 'onlineSavingsMsg'
Learn More

Cystatin A Antibody (1B12), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP2-59470UV

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213550

  • 562.62€ / 0.10 ml
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

Cystatin A Monoclonal antibody specifically detects Cystatin A in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Especificaciones

Especificaciones

Cystatin A
Monoclonal
DyLight 350
cystatin A (stefin A), cystatin AS, cystatin-A, STF1Cystatin-AS, STFAStefin-A
Recombinant human Cystatin A (1-98aa) purified from E. coli
0.1 mL
Cancer, Tumor Suppressors
1475
Store at 4°C in the dark.
IgG1 κ
Western Blot, Flow Cytometry, ELISA, Immunofluorescence
1B12
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales