missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CYP4A11 (Human) Recombinant Protein
Human CYP4A11 full-length ORF ( AAH22851, 1 a.a. - 215 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00001579-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MSVSVLSPSRLLGDVSGILQAASLLILLLLLIKAVQLYLHRQWLLKALQQSPCPPSHWLFGHIQELQQDQELQRIQKWVETFPSACPHWLWGGKVRVQLYDPDYMKVILGRSDPKSHGSYRFLAPWIGYGLLLLNGQTWFQHRRMLTPAFHYDILKPYVGLMADSVRVMLDKWEELLGQDSPLEVFQHVSLMTLDTIMKCAFRHWQRAQHSRHLP
Especificaciones
AAH22851 | |
cytochrome P450, family 4, subfamily A, polypeptide 11 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CYP4A11 | |
GST |
1579 | |
Wheat germ expression system | |
25 μg | |
CP4Y, CYP4A2, CYP4AII | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |