missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Cynomolgus LILRB4 / CD85k / ILT3 Protein, Fc Tag

539.84€ - 3335.05€

Especificaciones

Número de acceso XP_015297198.1
Peso molecular 52.7 kDa
Fuente Cynomolgus LILRB4, Fc Tag (CDK-C5258) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Glu 259 (Accession # XP_015297198.1).
Inmunógeno Gln 22 - Glu 259
Requisitos de almacenamiento -20°C
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
15806417
Ver documentos Detalles de la promoción
ACROBiosystems
CDK-C5258-100UG
100 μg
539.84€
100 microgramos
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 11-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
16477379
Ver documentos Detalles de la promoción
ACROBiosystems
CDK-C5258-1MG
1 mg
3335.05€
1 miligramo
Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío: 11-04-2024
para ver el stock
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Descripción

Descripción

Leukocyte immunoglobulin-like receptor subfamily B member 4 (LILRB4) is also known as CD85 antigen-like family member K (CD85K), Immunoglobulin-like transcript 3 (ILT-3), Leukocyte immunoglobulin-like receptor 5 (LIR-5), Monocyte inhibitory receptor HM18, which belongs to the leukocyte immunoglobulin-like receptor (LIR) family. LILRB4 / CD85K contains 2 Ig-like C2-type (immunoglobulin-like) domains. CD85K is detected in monocytes, macrophages, dendritic cells, lung, natural killer cells and B-cells. LILRB4 / CD85K is receptor for class I MHC antigens. CD85K recognizes a broad spectrum of HLA-A, HLA-B, HLA-C and HLA-G alleles, involved in the down-regulation of the immune response and the development of tolerance. LILRB4 interferes with TNFRSF5-signaling and NF-kappa-B up-regulation and inhibits receptor-mediated phosphorylation of cellular proteins and mobilization of intracellular calcium ions.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 52.7 kDa. As a result of glycosylation, the protein migrates as 60-66 kDa under reducing (R) condition, and 120-140 kDa under non-reducing (NR) condition (SDS-P
Especificaciones

Especificaciones

XP_015297198.1
Cynomolgus LILRB4, Fc Tag (CDK-C5258) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Glu 259 (Accession # XP_015297198.1).
-20°C
LILRB4
Cynomolgus Monkey
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Immune Checkpoint Proteins
52.7 kDa
Gln 22 - Glu 259
LILRB4,ILT3,LIR5,CD85K,HM18
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>85% as determined by SDS-PAGE
Videos
SDS
Documentos

Documentos

Product Certifications