Learn More
Cowie Technology™ Cylindrical PTFE Stir Bars
Product Code. 16245721 Shop All Fisherbrand Electrochemistry Products
Change view
Click to view available options
Length (Metric):
10 mm
12 mm
40 mm
Diameter (Metric):
4.5 mm
6 mm
8 mm
Unit Size:
Each
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Unit Size
16245721 10 mm 6 mm Pack of 10
10613161 12 mm 4.5 mm Each
10486671 40 mm 8 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16245721
Fecha estimada de envoi: 15-12-2026
Add to basket
Add to basket
Provides stable stirring action. Cowie Technology™ Cylindrical PTFE Stir Bars are a popular general-purpose stirrer for a wide variety of applications.
Provides stable stirring action. Cowie Technology™ Cylindrical PTFE Stir Bars are a popular general-purpose stirrer for a wide variety of applications.
  • Stable, general-purpose magnetic stir bar
  • Smooth, round profile for a wide variety of applications
  • Autoclavable
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Diameter (Metric) 6 mm
Length (Metric) 10 mm
Quantity 10/Pk.
Shape Cylinder