Learn More
KGW Isotherm™ Cylindrical Dewar Flasks, Blue with Handle
Product Code. 10187321
Change view
Click to view available options
Capacity (Metric):
1 L
1500 mL
Diameter (Metric) Outer:
95-105 mm
120-130 mm
Height (Metric):
290 mm
300 mm
Model:
G 15 C
G 9 C
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric) Outer Height (Metric) Model Unit Size
10187321 1500 mL 120-130 mm 300 mm G 15 C Each
15781977 1 L 95-105 mm 290 mm G 9 C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10187321 Supplier KGW Isotherm™ Supplier No. 10612
Fecha estimada de envoi: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Dewar flasks according to DIN EN ISO 16496. KGW-Isotherm Cylindrical Dewar Flasks, Blue with Side Handle provide high insulation reliability and easy handling in an economical product. Available in a variety of capacities, with plugs sold separately.
  • Made of borosilicate glass according to ISO 3585
  • Protective casing made of blue-coated metal
  • Fully silvered
  • Easy handling with side grip handle
  • Area of application:
    • Flexible laboratory, medical and biotechnology applications
    • Insulated storing and short-distance transport of small samples
    • Store under liquid or solid coolants (e.g. liquid N2 and CO2 dry ice)
    • Pressure-less space for coolants
    Chemical characteristics:
    • Hydrolytic resistance: according to ISO 719 (98°C)
    • Hydrolytic resistance: according to ISO 720 (121°C)
    • Acid resistance: according to ISO 1776
    • Alkaline resistance: according to ISO 695-A2
  • Temperature range:
    • Glass refill from -200°C to + 150°C, for a short time 200°C
    • Casing with mounting rubber and Latizell buffer from -10°C to + 60°C
Specifications
Height (Metric) 300 mm
Color Blue
Cap Lid Type Plug
Diameter (Metric) Outer 120-130 mm
Capacity (Metric) 1500 mL
Diameter (Metric) Inner 100 mm
For Use With (Application) Insulated storage and small distance transport
For Use With (Equipment) Coolant
Height (Metric) Inner 240 mm
Includes Handle/Side grip
Model G 15 C
Base Material Coated Metal
Material Borosilicate Glass
Style Fully silvered