missing translation for 'onlineSavingsMsg'
Learn More

Tetracarbonilo de ciclopentadienilvanadio, + 97 %, Thermo Scientific Chemicals

Catalog Number p-7080351
Click to view available options
Cantidad:
0.5 g
2 g
12108-04-2
C9H5O4V
228.076
MFCD00014259
LIFGJKDEFQAOFY-UHFFFAOYSA-N
carbon monoxide; cyclopentane; vanadium, tetracarbonyl .eta.5-cyclopenta-2,4-dien-1-yl vanadium
498652
monóxido de carbono; ciclopentano; vanadio
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[CH]1[CH][CH][CH][CH]1.[V]
12108-04-2
C9H5O4V
228.076
MFCD00014259
LIFGJKDEFQAOFY-UHFFFAOYSA-N
carbon monoxide; cyclopentane; vanadium, tetracarbonyl .eta.5-cyclopenta-2,4-dien-1-yl vanadium
498652
monóxido de carbono; ciclopentano; vanadio
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[CH]1[CH][CH][CH][CH]1.[V]

Employed as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Cyclopentadienylvanadium tetracarbonyl
Punto de fusión 138°C
Punto de ebullición 80°C to 100°C (0.1 mmHg)(sublimation)
Cantidad 0.5 g
Número UN UN3466
Sensibilidad Air Sensitive
Información de solubilidad Insoluble in water.
Formula Weight (peso de la fórmula) 228.08
Porcentaje de pureza ≥97%
Forma física cristalino/a/s

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'