missing translation for 'onlineSavingsMsg'
Learn More

Bromuro de ciclohexiltrifenilfosfonio, +98 %, Thermo Scientific Chemicals

Catalog Number 11442477
Click to view available options
Cantidad:
5 g
25 g
100 g
Este artículo no se puede devolver. Vea la política de devoluciones
7333-51-9
C24H26BrP
425.35
MFCD00031667
QRAKRDZMIONMRZ-UHFFFAOYSA-M
cyclohexyltriphenylphosphonium bromide, cyclohexyl triphenylphosphonium bromide, cyclohexyltriphenylphosphanium bromide, cyclohexyl triphenyl phosphanium bromide, acmc-20alcd, cyclohexyltriphenylphosphoniumbromide, cyclohexyl triphenyl phosphonium bromide
2724861
[Br-].C1CCC(CC1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Este artículo no se puede devolver. Vea la política de devoluciones
7333-51-9
C24H26BrP
425.35
MFCD00031667
QRAKRDZMIONMRZ-UHFFFAOYSA-M
cyclohexyltriphenylphosphonium bromide, cyclohexyl triphenylphosphonium bromide, cyclohexyltriphenylphosphanium bromide, cyclohexyl triphenyl phosphanium bromide, acmc-20alcd, cyclohexyltriphenylphosphoniumbromide, cyclohexyl triphenyl phosphonium bromide
2724861
[Br-].C1CCC(CC1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Cyclohexyltriphenylphosphonium bromide
Punto de fusión 265°C to 272°C
Cantidad 100 g
Beilstein 3581012
Sensibilidad Hygroscopic
Nombre IUPAC cyclohexyltriphenylphosphanium bromide
Formula Weight (peso de la fórmula) 425.36
Porcentaje de pureza ≥98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'