Learn More
Cyclo(-Gly-Phe), TRC
Product Code. 30367967
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Product Code. Quantity Unit Size
30367967 500 mg 500mg
1 options
This item is not returnable. View return policy
Product Code. 30367967 Supplier Toronto Research Chemicals Supplier No. TRCC989715500MG
 Restricted Item
Fecha estimada de envoi: 26-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Cyclo(-Gly-Phe)
CAS 10125-07-2
Molecular Formula C11H12N2O2
Quantity 500 mg
Synonym (S)-3-(Phenylmethyl)-2,5-piperazinedione, L-3-Benzyl-2,5-piperazinedione, Cyclo(glycyl-L-phenylalanyl), (S)-2-Benzylpiperazine-3,6-dione, (S)-3-Benzyl-2,5-dioxopiperazine, (S)-3-Benzylpiperazine-2,5-dione, Cyclic glycylphenylalanine, Cyclic(glycylphenylalanyl), Cyclo(Gly-L-Phe), Cyclo(L-phenylalanylglycine), Cyclo(L-phenylalanylglycyl), Cyclo-glycyl-L-phenylalanine
InChI Formula InChI=1S/C11H12N2O2/c14-10-7-12-11(15)9(13-10)6-8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H,12,15)(H,13,14)/t9-/m0/s1
SMILES O=C1CNC(=O)[C@H](Cc2ccccc2)N1
IUPAC Name (3S)-3-benzylpiperazine-2,5-dione
Molecular Weight (g/mol) 204.23
Formula Weight 204.0899