missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclin D3 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 26755-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The immunogen for the anti-h-Cyclin D3 was a recombit full length Cyclin D3 protein recognizing a 34 kD protein. Cyclin D3 is a G1 cyclin closedly related to Cyclins D1 and D2 phase progression. Like Cyclin D1 and Cyclin D2, it is activated by the same mechanisms and can phosphorylate pRB when associated with cdk4 and cdk6.Especificaciones
Cyclin D3 | |
Polyclonal | |
Unconjugated | |
CCND3 | |
CCND3, Cyclin D3, G1/S specific cyclin D3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25193, 896 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P30281, P48961 | |
CCND3 | |
Cyclin D3 Fusion Protein Ag25151 | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |