Learn More
Novus Biologicals™ Cyclin A1 Overexpression Lysate
Marca:  Novus Biologicals™ NBL1-08863
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18213393
  • 685.73€ / 0.10 miligramo
Fecha estimada de envío: 19-08-2024
para ver el stock
Descripción
Descripción
The Cyclin A1 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Especificaciones
Especificaciones
Western Blot
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
The lysate was created in HEK293T cells, using plasmid ID RC26615 and based on accession number NM_3914. The protein contains a C-terminal DDK tag.
RIPA buffer
Homo sapiens cyclin A1 (CCNA1), transcript variant 1, mRNA.
Native