missing translation for 'onlineSavingsMsg'
Learn More

CXCR1/IL-8RA Antibody (8F1-1-4), PE, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NBP1-43339PE

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18216032

  • 519.42€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

CXCR1/IL-8RA Monoclonal specifically detects CXCR1/IL-8RA in Human, Hamster samples. It is validated for Flow Cytometry.
Especificaciones

Especificaciones

CXCR1/IL-8RA
Monoclonal
PE
PBS with 0.05% Sodium Azide
CXCR1
The immunogen for this antibody was CD181.
0.1 mL
Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction
3577
Store at 4C in the dark.
IgG2b κ
Flow Cytometry
8F1-1-4
Flow Cytometry
CD128, CD181, CD181 antigen, CDw128aC-C, chemokine (C-X-C motif) receptor 1, CKR-1, CMKAR1, C-X-C chemokine receptor type 1, CXC-R1, CXCR-1, High affinity interleukin-8 receptor A, IL-8 receptor type 1, IL-8R A, IL8R1, IL8RAC-C-CKR-1, IL8RBA, interleukin 8 receptor, alpha, interleukin-8 receptor type 1, interleukin-8 receptor type A
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales