missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CXCL1 (Human) ELISA Kit

In-Vitro Enzyme-Linked Immunosorbent assay for the quantitative measurement of Human CXCL1

Marca:  Abnova™ KA1722.1Kit

Detalles adicionales : Peso : 0.02000kg

 Ver más versiones de este producto

Código de producto. 16056755

  • 578.00€ / 1 set


Fecha estimada de envío
11-12-2019
Añadir a la cesta

Especificaciones

Especificaciones

Biotin
CXCL1 (Human) ELISA Kit is an in vitro enzymelinked immunosorbent assay for the quantitative measurement of human CXCL1.
Colorimetric
96-well Antibody coated microwell, 25mL Wash Buffer 20X, 2 vials standard, 30mL assay diluent A, 15mL assay diluent B, 2 vials detection Antibody MSP, 8μL HRP Streptavidian concentrate, 12mL TMB One-Step Substrate reagent, 8mL stop solut
96 Assays
100μL
100μL
Human
50pg/mL
Quantification
CXCL1 (Human) ELISA Kit
Cell Culture Supernatant, Plasma, Serum, Urine
Store the kit at -20°C. Standard should be stored at -20°C or -80°C. Avoid repeated freezing and thawing.
Videos
Documentos