Learn More
Ambion™ Customer-Defined Pre-miR™ miRNA Precursor
Product Code. 15561154
Change view
Click to view available options
Product Type:
Pre-miRNA Precursor
Unit Size:
Each
Product Code. Product Type Unit Size
15561154 Pre-miRNA Precursor Each
1 options
This item is not returnable. View return policy
Product Code. 15561154 Supplier Ambion™ Supplier No. AM17103
 Restricted Item
Fecha estimada de envoi: 01-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Customer-Defined Pre-miR™ miRNA Precursor
Pre-miR™ miRNA Precursor Molecules are small, chemically modified double-stranded RNA molecules designed to mimic endogenous mature miRNAs. Use our customization tool to design the ideal mimic for your miRNA sequence, or select from among predesigned products.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Room temperature
Description Pre-miRNA Precursor, 20nmol, Small, Chemically Modified Double-Stranded RNA Molecules Designed to Mimic Endogenous Mature miRNAs, Use Our Customization Tool to Design the Ideal Mimic for Your miRNA Sequence
Product Type Pre-miRNA Precursor
Quantity 20 nmol
For Research Use Only. Not for use in diagnostic procedures.