Learn More
Ambion™ Custom Select siRNA
Custom Select siRNA
Marca:  Ambion™ 4390833
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15501471
  • 295.00€ / cada uno
Fecha de envío estimada
del proveedor 30-10-2019
Añadir a la cesta
Descripción
Descripción
Silencer™ Select custom siRNAs (LNA-modified 21-bp duplex with overhangs) are RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types. These siRNAs provide high knock-down efficiencies and specificities.
Especificaciones
Especificaciones
Custom Size Select siRNA, 6 to 8pH, Liquid, Ideal for Knocking Down Gene Expression in Order to Study Protein Function in a Wide Range of Cell Types, These siRNAs Provide High Knock-down Efficiencies and Specificities
Liquid
Custom Size Select siRNA
Knocking down gene expression in order to study protein function in a wide range of cell types
6 to 8
Room temperature
Videos
Documentos
For Research Use Only. Not for use in diagnostic procedures.