Learn More
Ambion™ Custom Pre-miR™ miRNA Precursor
Product Code. 10417114
Change view
Click to view available options
Content And Storage:
Room temperature
Unit Size:
Each
Product Code. Content And Storage Unit Size
10417114 Room temperature Each
1 options
This item is not returnable. View return policy
Product Code. 10417114 Supplier Ambion™ Supplier No. AM17101
 Restricted Item
Fecha estimada de envoi: 02-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Custom Pre-miR™ miRNA Precursor
miRNA Precursor Molecules are small, chemically modified double-stranded RNA molecules designed to mimic endogenous mature miRNAs.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Room temperature
Description Custom Anti-miR™ miRNA Precursor, 20nmol, Powder, miRNA Precursor Molecules are Small, Chemically Modified Double-stranded RNA Molecules Designed to Mimic Endogenous Mature miRNAs
Product Type Custom Anti-miR™ miRNA Precursor
Quantity 20 nmol
For Research Use Only. Not for use in diagnostic procedures.