missing translation for 'onlineSavingsMsg'
Learn More

Cupric Acetate, Monohydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog Number p-5390740
Click to view available options
Cantidad:
500 g
2.5 kg
Envase:
Frasco de polietileno
Poly Bottle
Este artículo no se puede devolver. Vea la política de devoluciones
6046-93-1
C4H8CuO5
199.65
MFCD00149570
NWFNSTOSIVLCJA-UHFFFAOYSA-L
copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
165397
copper;diacetate;hydrate
O.[Cu++].CC([O-])=O.CC([O-])=O
Este artículo no se puede devolver. Vea la política de devoluciones
6046-93-1
C4H8CuO5
199.65
MFCD00149570
NWFNSTOSIVLCJA-UHFFFAOYSA-L
copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
165397
copper;diacetate;hydrate
O.[Cu++].CC([O-])=O.CC([O-])=O

High quality chemicals for laboratory and specialized industrial use

Nombre del producto químico o material Cupric Acetate
Nombre de nota Monohydrate
Forma física Crystals
Cantidad 500 g
Formula Weight (peso de la fórmula) 199.65
Grado BAKER ANALYZED™ ACS Reagent
Envase Poly Bottle
Gravedad específica 1L = 1.88kg
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'