Learn More
Simport™ Cultube™ Sterile Culture Tubes, Polypropylene
For users who prefer a more compact package with tubes oriented horizontally.
Marca:  Simport™ T405-2A
Detalles adicionales : Peso : 0.01000kg
Código de producto. 11631460
  • 161.00€ / paquete de 500
Fecha estimada de envío
18-03-2020
Añadir a la cesta
Descripción
Descripción
  • Manufactured from either polystyrene or polypropylene
  • Tubes are placed in an convenient space-saving plastic tray
  • State-of-the-art packaging keeps your tubes neatly aligned for ease of manipulation
Especificaciones
Especificaciones
25/Tray; 500/Case
75 mm
Culture Test Tube
5 mL
Polypropylene
Sterile