Learn More
Sartorius™ Cubis™ Preconfigured Analytical Balance
Designed for weighing with maximum accuracy, without complex workflows.
Marca: Sartorius™ MSE224SCE-ED15
Detalles adicionales : Peso : 8.00000kg
Descripción
- Large, high-contrast LCD
- Simple menu guidance with short-text prompts
- Clear structured keys
Especificaciones
0.1 g | |
±0.07 mg | |
85 mm |
±0.2 mg | |
Preconfigured Analytical Balance | |
85 mm |