Learn More
Sartorius Cubis™ III Precision Balance, 1.5, 3.0, 6.2 kg, EU Verified Except France
Product Code. 31932749 Shop All Sartorius Products
Change view
Click to view available options
Readability:
10, 20, 50 mg
Capacity:
1.5, 3.0, 6.2 kg
Unit Size:
Each
Product Code. Readability Capacity Unit Size
31932749 10, 20, 50 mg 1.5, 3.0, 6.2 kg Each
1 options
This item is not returnable. View return policy
Product Code. 31932749 Supplier Sartorius Supplier No. CUB6202P1EU0
Fecha estimada de envoi: 13-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
High-performance precision balance engineered for accurate and reliable weighing in laboratory, industrial, and quality control applications.
  • Cubis III electronic precision balance with 7 in.
  • color TFT touch display
  • Actual scale division (d) 10, 20, 50 mg, nominal maximum load (Max) 1.5, 3.0, 6.2 kg
  • No draft shield
  • Verified balance with EC Type Approval Certificate (for EU except France)
  • Cables with plug types for EU, UK, Switzerland and Italy included
TRUSTED_SUSTAINABILITY
Specifications
Capacity 1.5, 3.0, 6.2 kg
Display Type Color TFT Touchscreen
Readability 10, 20, 50 mg
Type Precision Balance