missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CTSO (Human) Recombinant Protein
Human CTSO full-length ORF ( AAH49206, 24 a.a. - 321 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00001519-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: DSRAPFTPTWPRSREREAAAFRESLNRHRYLNSLFPSENSTAFYGINQFSYLFPEEFKAIYLRSKPSKFPRYSAEVHMSIPNVSLPLRFDWRDKQVVTQVRNQQMCGGCWAFSVVGAVESAYAIKGKPLEDLSVQQVIDCSYNNYGCNGGSTLNALNWLNKMQVKLVKDSEYPFKAQNGLCHYFSGSHSGFSIKGYSAYDFSDQEDEMAKALLTFGPLVVIVDAVSWQDYLGGIIQHHCSSGEANHAVLITGFDKTGSTPYWIVRNSWGSSWGVDGYAHVKMGSNVCGIADSVSSIFV
Especificaciones
AAH49206 | |
cathepsin O | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CTSO | |
GST |
1519 | |
Wheat germ expression system | |
25 μg | |
CTSO1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |