Learn More
Ctip1 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca:  Bio-Techne NB600-261FR
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18215911
  • 620.93€ / 100 microlitros
Fecha estimada de envío: 21-08-2024
para ver el stock
Descripción
Descripción
Ctip1 Polyclonal specifically detects Ctip1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.
Especificaciones
Especificaciones
Ctip1
Polyclonal
Western Blot, Flow Cytometry
B-cell CLL/lymphoma 11A, B-cell CLL/lymphoma 11A (zinc finger protein), B-cell CLL/lymphoma 11A (zinc finger protein) isoform 2, B-cell lymphoma/leukemia 11A, BCL-11A, BCL11A B-cell CLL/lymphoma 11A (zinc finger protein) isoform 1, BCL11A-S, BCL11A-XL, C2H2-type zinc finger protein, COUP-TF-interacting protein 1, CTIP1ecotropic viral integration site 9, ecotropic viral integration site 9 homolog, Ecotropic viral integration site 9 protein homolog, EVI-9, EVI9FLJ34997, HBFQTL5, KIAA1809BCL11A-L, Zinc finger protein 856, ZNF856FLJ10173
Rabbit
Affinity Purified
RUO
Primary
NB600-261 is specific for human Bcl-11a protein.
Store at 4C in the dark.
Western Blot, Flow Cytometry
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
BCL11A
A synthetic peptide which maps to a region between residues 775 and the C-terminus (residue 835) of human B-cell CLL/Lymphoma 11A using the numbering given in Swiss-Prot entry Q9H165 (GeneID 53335).
100 μL
Apoptosis, Cancer, Inhibitors Activators and Regulators, Transcription Factors and Regulators, Tumor Suppressors
53335
Human, Mouse