missing translation for 'onlineSavingsMsg'
Learn More

CTHRC1 Rabbit anti-Human, Clone: 166, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Novus Biologicals NBP2-89947-100UL

Detalles adicionales : Peso : 0.00970kg

 Ver más versiones de este producto

Código de producto. 18214447

  • 537.78€ / 100 microlitros
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

CTHRC1 Monoclonal specifically detects CTHRC1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Especificaciones

Especificaciones

CTHRC1
Monoclonal
Unconjugated
0.2 um filtered solution in PBS
Rabbit
Protein A purified
RUO
115908
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunoprecipitation
166
Western Blot 1:500-1:2000, Immunoprecipitation 1-4 ul/mg of lysate
collagen triple helix repeat containing 1, collagen triple helix repeat-containing protein 1, Protein NMTC1
Recombinant Human CTHRC1 protein
100 μg
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales