missing translation for 'onlineSavingsMsg'
Learn More

CTAG1B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Bio-Techne NBP2-88796

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18215547

  • 455.71€ / 100 microlitros
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

CTAG1B Polyclonal specifically detects CTAG1B in Human samples. It is validated for Western Blot.
Especificaciones

Especificaciones

CTAG1B
Polyclonal
Unconjugated
PBS, 2% Sucrose
Rabbit
Affinity purified
RUO
1485
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
Autoimmunogenic cancer/testis antigen NY-ESO-1, cancer antigen 3, cancer/testis antigen 1, cancer/testis antigen 1B, Cancer/testis antigen 6.1, CT6.1L antigen family member 2, CTAG1CTAG1A, ESO1CTAG, LAGE2, LAGE2A, LAGE2BLAGE-2, New York esophageal squamous cell carcinoma 1, NY-ESO-1
The immunogen is a synthetic peptide directed towards the C-terminal region of human CTAG1B
100 μL
Primary
Human
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales