Learn More
CleanSpace™ Technology CST Neck Support
Product Code. 17845085 Shop All CleanSpace Technology Products
Change view
Click to view available options
Size:
Small
Medium
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17845085 Small Each
17805095 Medium Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17845085 Supplier CleanSpace™ Technology Supplier No. CST1019
  • ONLINE EXCLUSIVE PRICE
    64.49€
    50.00€ / paquete de 1
    Ahorro 14.49€
    22% Descuento
Fecha estimada de envoi: 13-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
CleanSpace CST Neck Supports are specifically designed to provide additional positioning options of the power unit for optimal wearer comfort.
  • CleanSpace CST Neck Supports are specifically designed to provide additional positioning options of the power unit for optimal wearer comfort.
  • Compatible with the CleanSpace PRO Power Unit (CST1002) and CleanSpace ULTRA Power Unit (CST1012).
  • Made of light weight polypropylene plastic.
  • Clips directly into the Power Unit and is easily changed.
  • A CleanSpace CST Neck Support is NOT certified intrinsically safe.
Specifications
Color Black
Disposable Reusable
Material Polypropylene
Product Type Neck Support
Size Small