Learn More
CleanSpace™ Technology CST Harness
Product Code. 17815095 Shop All CleanSpace Technology Products
Change view
Click to view available options
Size:
One Size
Unit Size:
Each
Product Code. Size Unit Size
17815095 One Size Each
1 options
This item is not returnable. View return policy
Product Code. 17815095 Supplier CleanSpace™ Technology Supplier No. CST1021
  • ONLINE EXCLUSIVE PRICE
    80.57€
    62.50€ / paquete de 1
    Ahorro 18.07€
    22% Descuento
Fecha estimada de envoi: 13-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
The CST Head Harness is specifically designed to aid in supporting the Power Unit for optimal comfort.
  • Specifically designed to provide additional positioning options of the power unit for optimal wearer comfort.
  • Compatible with the CleanSpace PRO Power Unit (CST1002) and CleanSpace ULTRA Power Unit (CST1012).
  • Made of light weight polypropylene plastic.
  • Clips directly into the Power Unit and is easily changed.
  • A CleanSpace CST Neck Support is NOT certified intrinsically safe.
Specifications
Color Black
Disposable Reusable
Material HDPE, Nylon, Silicone
Product Type Head Harness
Size One Size