Learn More
CleanSpace™ Technology CST Half Mask with Head Harness
Product Code. 17840705 Shop All CleanSpace Technology Products
Change view
Click to view available options
Size:
Large/X-Large
Medium
Small
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17840705 Medium Each
17825085 Small Each
17835085 Large/X-Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17840705 Supplier CleanSpace™ Technology Supplier No. CST1035
  • ONLINE EXCLUSIVE PRICE
    179.99€
    139.00€ / paquete de 1
    Ahorro 40.99€
    22% Descuento
Fecha estimada de envoi: 05-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
A CleanSpace CST Half Masks (including a CleanSpace CST Head Harness) is designed for use with the CleanSpace PRO Power Unit (CST1000) and CleanSpace ULTRA Power Unit (CST 1010).
  • Designed for use with the CleanSpace PRO Power Unit (CST1000) and CleanSpace ULTRA Power Unit (CST 1010).
  • Made of soft silicone that readily moulds to the face and does not cause allergic dermatitis.
  • The easy-to-locate clips secure the CleanSpace CST Half Mask to the Power Unit.
  • Attaches easily to connection points on the mask arms and can be removed for cleaning.
  • A CleanSpace CST Half Mask (including a CleanSpace CST Head Harness) is NOT certified intrinsically safe.
Specifications
Color Black
Disposable Reusable
Material Silicon
Product Type 1/2 Mask and Harness
Size Medium