missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CSRP2 (Human) Recombinant Protein
Human CSRP2 full-length ORF ( AAH00992, 1 a.a. - 193 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00001466-P01.25ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MPVWGGGNKCGACGRTVYHAEEVQCDGRSFHRCCFLCMVCRKNLDSTTVAIHDEEIYCKSCYGKKYGPKGYGYGQGAGTLNMDRGERLGIKPESVQPHRPTTNPNTSKFAQKYGGAEKCSRCGDSVYAAEKIIGAGKPWHKNCFRCAKCGKSLESTTLTEKEGEIYCKGCYAKNFGPKGFGYGQGAGALVHAQ
Especificaciones
AAH00992 | |
cysteine and glycine-rich protein 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CSRP2 | |
GST |
1466 | |
Wheat germ expression system | |
25 μg | |
CRP2, LMO5, SmLIM | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |