missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CSNK2A1 (CK2 α 1) Recombinant Human Protein
CSNK2A1 is a serine/threonine protein kinase.
Marca: Invitrogen™ PR4554A
Detalles adicionales : Peso : 0.01000kg
Descripción
CSNK2A1 phosphorylates acidic proteins.
Especificaciones
NP_001883.4 | |
50mM Tris, pH 7.5, 100mM NaCl, 0.05mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
CSNK2A1 (CK2 α 1) | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
47.9kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
CK2 Kinase Family | |
Recombinant | |
Baculovirus |