Learn More
Invitrogen™ CSNK1G2 (CK1 γ 2) Recombinant Human Protein
CSNK1G2 (CK1 gamma 2) is a serine/threonine kinase.
Marca: Invitrogen™ PR5511A
Detalles adicionales : Peso : 0.01000kg
Descripción
CSNK1G2 (CK1 gamma 2) is implicated in a wide range of cellular functions, including vesicular trafficking, DNA damage repair, cell cycle progression, and cytokinesis.
Especificaciones
NP_001310 | |
50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.04% Triton X-100, 4mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
CSNK1G2 (CK1 γ 2) | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
52.5kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
CK1 Kinase Family | |
Recombinant | |
Baculovirus |