missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Human CSK, His Tag Recombinant Protein
Human CSK, His Tag Recombinant Protein
Marca: Thermo Scientific™ P2927
Detalles adicionales : Peso : 3.36000kg
Descripción
CSK is a tyrosine kinase that has been shown to downregulate the activity of SRC oncoprotein through phosphorylation of its carboxy terminus. Since cell transformation by SRC is caused by various mechanisms that interfere with this phosphorylation, the CSK gene might function as an antioncogene.Especificaciones
NP_004374 | |
1445 | |
10 μg | |
CSK Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris, pH 7.5, 100 mM NaCl, 10% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Tyrosine Kinases | |
LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™ | |
c-src tyrosine kinase | |
Kinases & Inhibitors | |
Cytoplasmic Tyrosine Kinases |
Kinase Assay | |
51.9 kDa | |
CSK | |
Human | |
His Tag | |
Biochemical binding/kinetic, Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity | |
Dry Ice | |
CSK | |
E. coli | |
Full Length, Recombinant, Cytoplasmic | |
Signal Transduction |