Learn More
CSK Antibody (CSK-04), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Marca:  Novus Biologicals NB500-344AF700
Detalles adicionales : Peso : 0.00970kg
Código de producto. 18214460
  • 497.83€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock
Descripción
Descripción
CSK Monoclonal specifically detects CSK in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Especificaciones
Especificaciones
CSK
Monoclonal
Alexa Fluor 700
C-Src kinase, c-src tyrosine kinase, EC 2.7.10, EC 2.7.10.2, MGC117393, Protein-tyrosine kinase CYL, tyrosine-protein kinase CSK
Mouse
Protein A purified
RUO
Primary
The antibody CSK-04 reacts with Csk (C-terminal Src kinase), a 50 kDa non-receptor protein tyrosine kinase expressed in cytosol.
Store at 4C in the dark.
IgG1
Western Blot, Immunocytochemistry, Immunofluorescence
CSK-04
Western Blot, Immunocytochemistry/Immunofluorescence
CSK
Bacterially expressed recombinant fragment of human Csk corresponding to amino acids 330-450.
0.1 mL
Cell Cycle and Replication, Signal Transduction
1445
Human, Mouse
Purified