missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSF1R (FMS) Recombinant Human Protein
CSF1R (FMS) Recombinant Human Protein
Marca: Invitrogen PV3249
Detalles adicionales : Peso : 3.36000kg
Descripción
CSF1R (FMS) is involved in the production, differentiation, and function of macrophages, and is implicated in promyelocytic leukemias.Especificación
NP_005202 | |
Pharma and biopharma, protein assays and analysis, target and lead identification and validation | |
44.8kDa | |
80% by SDS-PAGE | |
–80°C | |
1 Each | |
CSF1R (FMS), FMS, CSFR, c-FMS, CD115 | |
CSF1R was pre-diluted in enzyme dilution buffer and assayed in 50mM HEPES (pH 7.5), 10mM MgCl2, 10% glycerol, 2.5mM DTT, 0.01% Triton™ X-100, 200μM ATP, 200μg/mL poly [Glu, Tyr] 4:1 substrate and trace [32P]-γ-ATP for 10 minutes at 30°C | |
Kinase and inhibitors | |
PDGFR | |
CSF1R (FMS) Recombinant Human Protein | |
Human |
0.36mg/mL | |
1436 | |
CSF1R (FMS) Recombinant Human Protein | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
Receptor tyrosine kinases | |
CSF1R (FMS) Recombinant Human Protein | |
Kinase assay | |
Baculovirus | |
Amino acids 538-910 | |
310nmoles of phosphate transferred to poly [Glu, Tyr] 4:1 substrate per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 2μg/mL | |
His Tag |
For Research Use Only. Not for use in diagnostic procedures.