Learn More
Abnova™ CSF1 (Human) Recombinant Protein
Human CSF1 (159 a.a.) full-length recombinant protein with His tag expressed in Barley grain (Hordeum vulgare).
Marca:  Abnova™ P5818.10ug
Detalles adicionales : Peso : 0.00010kg
 Ver más versiones de este producto
Código de producto. 16202300
  • 120.00€ / 10 microgramos
Fecha estimada de envío
23-10-2020
Añadir a la cesta
Especificaciones
Especificaciones
BAD92189
Lyophilized
colony stimulating factor 1 (macrophage)
SDS-PAGE Stained with Coomassie Blue
Store at -20°C on dry atmosphere. After reconstitution with sterile water to a concentration of no less than 100 ug/mL, store at -20°C. For long term storage it is recommended to add a carrier protein (0.1% HSA or BSA). Aliquot to avoid repeated freezing and thawing.
CSF1
His
100 μg/mL
1435
Barley grain (Hordeum vulgare) expression system
10 μg
MCSF, MGC31930
Plants
Lyophilized from 5 mM TrisHCl, 80 mM Arginine, pH 8.0